Please login to your online account to display your discounted pricing

Tris-EDTA, 100X Solution (Molecular Biology), Fisher BioReagents


Quantity 1L
Packaging Nalgene Poly Bottle

Manufacturer: fisher scientific company  V36702

 View more versions of this product

Catalog No. BP1338-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 1L
Synonym TE
Filtered Through Filtered through a 0.2-micron filter.
Color Clear
Physical Form Liquid
DNase DNase free
Protease Protease free
Chemical Name or Material Tris-EDTA
Name Note 100X Solution
CAS 77-86-1
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Health Hazard 2 WARNING!
CAS 60-00-4
Packaging Nalgene Poly Bottle
Recommended Storage RT
CAS 7732-18-5

Specifications

Chemical Name or Material Tris-EDTA
Name Note 100X Solution
CAS 77-86-1
CAS 60-00-4
CAS 7732-18-5
Purity Grade Molecular Biology Grade

  • Simplify your buffer prepraration by using ready-made Fisher BioReagents 100X concentrate TE buffer solution -simply dilute as needed
  • 100X solution contains 1.0M tris and 0.1M EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer (pH 8.0) is routinely used for resuspending DNA after precipitation
  • Intended for laboratory research use only

Accessories
 

Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

RT

SDS & Safety Documents