Please login to your online account to display your discounted pricing

Tris-EDTA, 100X Solution (Molecular Biology), Fisher BioReagents

$119.49 - $119.49

Chemical Identifiers

Chemical Name or Material Tris-EDTA
Name Note 100X Solution
CAS 77-86-1
CAS 60-00-4
CAS 7732-18-5
Purity Grade Molecular Biology Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP1338-1

  SDS
fisher scientific company
V36702
Nalgene Poly Bottle 1L Each for $119.49
Case of 6 Each for $716.94
Description & Specifications

Specifications

Purity Grade Molecular Biology Grade
Quantity 1L
Synonym TE
Filtered Through Filtered through a 0.2-micron filter.
Color Clear
Physical Form Liquid
DNase DNase free
Protease Protease free
Chemical Name or Material Tris-EDTA
Name Note 100X Solution
CAS 77-86-1
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Health Hazard 2 WARNING!
CAS 60-00-4
Packaging Nalgene Poly Bottle
Recommended Storage RT
CAS 7732-18-5

Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.

Safety and Handling
 

Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

RT