missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Dodecyl Sulfate [for Biochemical Research], TCI America™

Manufacturer:  TCI America T30711G

 View more versions of this product

Catalog No. T30711G



Specifications

Specifications

Tris Dodecyl Sulfate [for Biochemical Research]
1g
C16H37NO7S
Lauryl Sulfate Tris Salt, Dodecyl Sulfate Tris(hydroxymethyl)aminomethane Salt
CCCCCCCCCCCCOS(=O)(=O)O.C(C(CO)(CO)N)O
387.532
387.53
≥96.0% (T)
White-Yellow
50996-85-5
MFCD00043277
TYTUELFFKLZOTE-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;dodecyl hydrogen sulfate
11703755
Crystal-Powder at 20°C

Chemical Identifiers

50996-85-5
387.532
TYTUELFFKLZOTE-UHFFFAOYSA-N
11703755
CCCCCCCCCCCCOS(=O)(=O)O.C(C(CO)(CO)N)O
C16H37NO7S
MFCD00043277
Lauryl Sulfate Tris Salt, Dodecyl Sulfate Tris(hydroxymethyl)aminomethane Salt
2-amino-2-(hydroxymethyl)propane-1,3-diol;dodecyl hydrogen sulfate
Safety and Handling

Safety and Handling

TSCA : No

Documents