missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(dioxa-3,6-heptyl)amine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 124851000
Specifications
Tris(dioxa-3, 6-heptyl)amine | |
162°C | |
Glass bottle | |
1.01 | |
Brown to Yellow | |
70384-51-9 | |
C15H33NO6 | |
MFCD00010748 | |
2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, tda-1, tris 2-2-methoxyethoxy ethyl amine, tris 3,6-dioxaheptyl amine, tris dioxa-3,6-heptyl amine, tris-2-2-methoxyethoxy ethyl amine | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine | |
112414 | |
Liquid |
1.0100g/mL | |
Authentic | |
1.4466 to 1.4486 | |
330.0°C | |
100mL | |
95% | |
(CH3OCH2CH2OCH2CH2)3N | |
15,356; 16,373 | |
Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane | |
COCCOCCN(CCOCCOC)CCOCCOC | |
323.42 | |
323.42 | |
95% |
Chemical Identifiers
70384-51-9 | |
323.42 | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
112414 | |
COCCOCCN(CCOCCOC)CCOCCOC |
C15H33NO6 | |
MFCD00010748 | |
2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, tda-1, tris 2-2-methoxyethoxy ethyl amine, tris 3,6-dioxaheptyl amine, tris dioxa-3,6-heptyl amine, tris-2-2-methoxyethoxy ethyl amine | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 274-590-5