missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(dioxa-3,6-heptyl)amine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC124850250

 View more versions of this product

Catalog No. AC124850250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Tris(dioxa-3, 6-heptyl)amine
162°C
Glass bottle
1.01
Yellow to Brown
70384-51-9
C15H33NO6
MFCD00010748
2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, tda-1, tris 2-2-methoxyethoxy ethyl amine, tris 3,6-dioxaheptyl amine, tris dioxa-3,6-heptyl amine, tris-2-2-methoxyethoxy ethyl amine
XGLVDUUYFKXKPL-UHFFFAOYSA-N
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
112414
Liquid
1.0100g/mL
Authentic
1.4466 to 1.4486
330.0°C
25mL
95%
(CH3OCH2CH2OCH2CH2)3N
15,356; 16,373
Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane
COCCOCCN(CCOCCOC)CCOCCOC
323.42
323.42
95%

Chemical Identifiers

70384-51-9
323.42
XGLVDUUYFKXKPL-UHFFFAOYSA-N
112414
COCCOCCN(CCOCCOC)CCOCCOC
C15H33NO6
MFCD00010748
2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, tda-1, tris 2-2-methoxyethoxy ethyl amine, tris 3,6-dioxaheptyl amine, tris dioxa-3,6-heptyl amine, tris-2-2-methoxyethoxy ethyl amine
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 274-590-5

SDS Documents