Please login to your online account to display your discounted pricing

Tris(dimethylamino)methane, 85%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tris(dimethylamino)methane
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C7H19N3
Linear Formula CH[N(CH3)2]3
Formula Weight 145.25
MDL Number MFCD00008322
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205700010

  SDS
thermo fisher scientific
AC205700010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205700050

  SDS
thermo fisher scientific
AC205700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥80%
Physical Form Liquid
Formula Weight 145.25
Quantity 1g
Color Clear colorless to slightly yellow
Density 0.8380g/mL
Chemical Name or Material Tris(dimethylamino)methane
Assay Percent Range 85%
Molecular Formula C7H19N3
Linear Formula CH[N(CH3)2]3
MDL Number MFCD00008322
CAS 5762-56-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Extremely flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Extremely flammable liquid and vapor.