Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(dimethylamino)gallium(III) dimer, 99.9% (metals basis)

Gallium nitride film precursor

Chemical Identifiers

Chemical Name or Material Tris(dimethylamino)gallium(III) dimer
CAS 57731-40-5
Linear Formula Ga2[N(CH3)2]6
Formula Weight 403.9
MDL Number MFCD00798535
UN Number UN3131
Synonym Gallium dimethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4044327

  SDS
alfa aesar™
4044327
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40443ME

  SDS
alfa aesar™
40443ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder/Chunks
Formula Weight 403.9
Packaging Ampule
Chemical Name or Material Tris(dimethylamino)gallium(III) dimer
Melting Point 91°C
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula Ga2[N(CH3)2]6
MDL Number MFCD00798535
CAS 57731-40-5
UN Number UN3131
Synonym Gallium dimethylamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
Color White to yellow
Boiling Point 125°C (0.01mmHg) (subl.)
Sensitivity Moisture Sensitive
Quantity 2.5g

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon