Please login to your online account to display your discounted pricing

Tris(diformylamino)methane, 98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380500050

 View more versions of this product

Catalog No. AC380500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.15
Physical Form Crystalline Powder
Quantity 5g
Color White
Infrared Spectrum Authentic
Linear Formula CH[N(CHO)2]3
Elemental Analysis 97.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H7N3O6
MDL Number MFCD08059357
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 98%
Packaging Glass bottle
Chemical Name or Material Tris(diformylamino)methane
CAS 332047-72-0

Specifications

Chemical Name or Material Tris(diformylamino)methane
Assay Percent Range 98%
Molecular Formula C7H7N3O6
Linear Formula CH[N(CHO)2]3
Formula Weight 229.15
MDL Number MFCD08059357
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents