Please login to your online account to display your discounted pricing

Tris(diformylamino)methane, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material Tris(diformylamino)methane
Assay Percent Range 98%
Molecular Formula C7H7N3O6
Linear Formula CH[N(CHO)2]3
Formula Weight 229.15
MDL Number MFCD08059357
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380500010

  SDS
thermo fisher scientific
AC380500010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380500050

  SDS
thermo fisher scientific
AC380500050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.15
Physical Form Crystalline Powder
Quantity 1g
Color White
Linear Formula CH[N(CHO)2]3
Elemental Analysis 97.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 332047-72-0
Packaging Glass bottle
Molecular Formula C7H7N3O6
MDL Number MFCD08059357
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 98%
Chemical Name or Material Tris(diformylamino)methane
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.