Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(diethylamino)gallium(III), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Tris (diethylamino)gallium (III)
CAS 194611-64-8
Molecular Formula C12H30GaN3
Linear Formula Ga[N(CH2CH3)2]3
Formula Weight 286.11
MDL Number MFCD01863475
UN Number UN3263
Synonym Gallium diethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4275909

  SDS
alfa aesar™
4275909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.11
Linear Formula Ga[N(CH2CH3)2]3
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Physical Form Powder
Health Hazard 1 Danger
Quantity 10g
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
Molecular Formula C12H30GaN3
UN Number UN3263
MDL Number MFCD01863475
Synonym Gallium diethylamide
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
Color Pale brown
Assay 99.9%
Chemical Name or Material Tris (diethylamino)gallium (III)
CAS 194611-64-8
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.