Please login to your online account to display your discounted pricing

Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369340050

 View more versions of this product

Catalog No. AC369340050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 1035.11
Physical Form Crystalline Powder
Assay ≥96.0% (ex Pd)
Quantity 5g
Color Purple to black
Fieser 14,339; 15,353; 16,372; 17,394
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Assay Percent Range 97%
Packaging Glass bottle
Synonym Dipalladium(0)tris(dibenzylideneacetone)-chloroform adduct
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2·CHCl3

Specifications

Chemical Name or Material Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Assay ≥96.0% (ex Pd)
Assay Percent Range 97%
Molecular Formula C52H43Cl3O3Pd2
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2·CHCl3
Formula Weight 1035.11
Synonym Dipalladium(0)tris(dibenzylideneacetone)-chloroform adduct
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.