Please login to your online account to display your discounted pricing

Tris(dibenzylideneacetone)dipalladium(0), Pd 21.5% min., Alfa Aesar™

$101.14 - $310.93

Chemical Identifiers

Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)
CAS 51364-51-3
Physical Form Powder
Molecular Formula C51H42O3Pd2
Formula Weight 915.74
MDL Number MFCD00013310
Synonym Bis[tris(dibenzylideneacetone)palladium(0)]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1276003

 
alfa aesar
1276003
1g Each for $101.14

AA1276006

 
alfa aesar
1276006
5g Each for $310.93
Description & Specifications

Specifications

Formula Weight 915.74
Quantity 1g
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)
Physical Form Powder
Molecular Formula C51H42O3Pd2
MDL Number MFCD00013310
CAS 51364-51-3
Synonym Bis[tris(dibenzylideneacetone)palladium(0)]