missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%

Manufacturer:  Alfa Aesar™ 4496903

 View more versions of this product

Catalog No. AA4496903



Specifications

Specifications

Tris (dibenzylideneacetone)dipalladium (0)
131°C to 135°C
Complex with chloroform
C52H43Cl3O3Pd2
MFCD00075479
Pd 20.6%
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C(Cl)(Cl)Cl.[Pd].[Pd]
1035.103
1035.11
Air & Moisture Sensitive
1g
52522-40-4
Pd2(C17H14O)3·CHCl3
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex, tris dibenzylideneacetone dipalladium 0 chloroform adduct
LNAMMBFJMYMQTO-FNEBRGMMSA-N
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
11029508
Powder

Chemical Identifiers

52522-40-4
1035.103
LNAMMBFJMYMQTO-FNEBRGMMSA-N
11029508
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C(Cl)(Cl)Cl.[Pd].[Pd]
C52H43Cl3O3Pd2
MFCD00075479
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex, tris dibenzylideneacetone dipalladium 0 chloroform adduct
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
Safety and Handling

Safety and Handling

GHS H Statement
H351-H373-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.

GHS P Statement
P260-P280-P281-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents