Learn More
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 4496906
Description
Tris(dibenzylideneacetone)dipalladium(0) is used in Synthesis of nanosized palladium phosphides upon interaction with white phosphorous, Preparation of palladium triphenylphosphine carbonyl cluster complexes, Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tris (dibenzylideneacetone)dipalladium (0) | |
52522-40-4 | |
C52H43Cl3O3Pd2 | |
MFCD00075479 | |
Air and moisture sensitive | |
Soluble in organic solvents. Insoluble in water. | |
[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1 | |
1035.10 | |
1035.11 | |
Powder |
Complex with chloroform | |
131°C to 135°C | |
Pd2(C17H14O)3·CHCl3 | |
5 g | |
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex | |
LNAMMBFJMYMQTO-UHFFFAOYSA-N | |
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium | |
11029508 | |
Pd 20.6% |
Chemical Identifiers
52522-40-4 | |
1035.10 | |
LNAMMBFJMYMQTO-UHFFFAOYSA-N | |
11029508 | |
[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1 |
C52H43Cl3O3Pd2 | |
MFCD00075479 | |
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex | |
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium |
Safety and Handling
GHS H Statement
H351-H373-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H320-H351
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only