Please login to your online account to display your discounted pricing

Tris DBA, Tocris™

N-myristoyltransferase-1 inhibitor; antiproliferative

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Tris DBA
Assay Percent Range >98%
Molecular Formula C51H42O3Pd2
Formula Weight 915.73
Synonym Tris(dibenzylideneacetone) dipalladium
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

341650

 
r & d systems
RDS341650
50mg Each for $546.19

341610

 
r & d systems
RDS341610
10mg Each for $129.05
Description & Specifications

Specifications

Formula Weight 915.73
Quantity 50mg
Chemical Name or Material Tris DBA
Assay Percent Range >98%
Molecular Formula C51H42O3Pd2
CAS 51364-51-3
Synonym Tris(dibenzylideneacetone) dipalladium
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature