missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(cyclopentadienyl)samarium(III), 98%

Manufacturer:  Alfa Aesar™ 4441106

 View more versions of this product

Catalog No. AA4441106



Description

Description

  • Ampouled under argon
Specifications

Specifications

Tris (cyclopentadienyl)samarium (III)
Air and moisture sensitive
5g
C15H15Sm
UN3181
HLLVKWMULLMCMH-UHFFFAOYSA-N
345.64
98%
Ampule
365°C
1298-55-1
MFCD00013755
samarium 3+ ion tris cyclopenta-1,3-dien-1-ide
345.65
Powder

Chemical Identifiers

1298-55-1
345.65
HLLVKWMULLMCMH-UHFFFAOYSA-N
C15H15Sm
MFCD00013755
samarium 3+ ion tris cyclopenta-1,3-dien-1-ide
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

EINECSNumber : 215-075-7

TSCA : No

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents