Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(cyclopentadienyl)lanthanum(III), 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Tris(cyclopentadienyl)lanthanum(III)
CAS 1272-23-7
Formula Weight 334.19
MDL Number MFCD00045467
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3925103

  SDS
alfa aesar™
3925103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3925106

  SDS
alfa aesar™
3925106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.19
Packaging Ampule
Chemical Name or Material Tris(cyclopentadienyl)lanthanum(III)
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00045467
CAS 1272-23-7
UN Number UN3181
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Melting Point 295°C (dec.)
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Sensitivity Air & Moisture Sensitive
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.

  • Ampouled under argon