missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(cyclopentadienyl)holmium(III), 98%

Manufacturer:  Alfa Aesar™ 4430203

 View more versions of this product

Catalog No. AA4430203



Description

Description

  • Ampouled under argon
Specifications

Specifications

Tris (cyclopentadienyl)holmium (III)
Moisture sensitive
295°C
1272-22-6
MFCD03427140
acmc-1bt45
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Ho+3]
360.215
360.21
98%
Ampule
230°C (sublimation)(vac.)
1g
C15H15Ho
UN3181
ZZSQGMHUOVLCBI-UHFFFAOYSA-N
cyclopenta-1,3-diene;holmium(3+)
21115617
Crystalline,Ampouled under argon

Chemical Identifiers

1272-22-6
360.215
ZZSQGMHUOVLCBI-UHFFFAOYSA-N
21115617
C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Ho+3]
C15H15Ho
MFCD03427140
acmc-1bt45
cyclopenta-1,3-diene;holmium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

EINECSNumber : 215-047-4

TSCA : No

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents