Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(cyclopentadienyl)holmium(III), 98%

Chemical Identifiers

Chemical Name or Material Tris (cyclopentadienyl)holmium (III)
CAS 1272-22-6
Molecular Formula C15H15Ho
Formula Weight 360.21
MDL Number MFCD03427140
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4430203

 
alfa aesar™
4430203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4430206

 
alfa aesar™
4430206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 360.21
Packaging Ampule
Chemical Name or Material Tris (cyclopentadienyl)holmium (III)
Physical Form Crystalline
Assay 98%
Molecular Formula C15H15Ho
MDL Number MFCD03427140
CAS 1272-22-6
UN Number UN3181
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Melting Point 295°C
Boiling Point 230°C (subl.) (vac.)
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Sensitivity Moisture Sensitive
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.

  • Ampouled under argon