Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(cyclopentadienyl)erbium(III), 99% (99.9%-Er) (REO)

Chemical Identifiers

Chemical Name or Material Tris(cyclopentadienyl)erbium(III)
CAS 39330-74-0
Name Note (REO)
Molecular Formula C15H15Er
Formula Weight 362.54
MDL Number MFCD00145475
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2738903

  SDS
alfa aesar™
H2738903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2738906

  SDS
alfa aesar™
H2738906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 362.54
Chemical Name or Material Tris(cyclopentadienyl)erbium(III)
Name Note (REO)
Assay 99%
Assay Percent Notes (99.9%-Er)
Molecular Formula C15H15Er
MDL Number MFCD00145475
CAS 39330-74-0
UN Number UN3181
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Melting Point 287°C
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.