missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS-buffered saline (TBS, 5X), with EDTA

$69.85 - $69.85

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63692K2 View Documents Alfa Aesar™
J63692K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ63692K3 View Documents Alfa Aesar™
J63692K3
2L Each for $69.85
 
Specifications

Chemical Identifiers

77-86-1
121.136
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

77-86-1
5X
C(C(CO)(CO)N)O
121.136
CHEBI:9754
1L
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Liquid
TRIS-buffered saline, with EDTA
Safety and Handling

Safety and Handling

TSCA : TSCA