missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS-buffered saline (TBS, 10X) pH 8.0

$61.38 - $171.18

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62780K2 View Documents Alfa Aesar™
J62780K2
1L Each for $61.38
 
AAJ62780K3 View Documents Alfa Aesar™
J62780K3
2L Each for $110.41
 
AAJ62780K7 View Documents Alfa Aesar™
J62780K7
4L Each for $171.18
 
Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Liquid
8.0
TRIS-buffered saline
C4H11NO3
10X
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Odorless
1L
Safety and Handling

Safety and Handling

TSCA : TSCA