missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS-buffered saline (TBS, 10X, low salt) pH 8.0

$51.04 - $165.01

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAJ60102K2 missing translation for 'viewDocuments' Alfa Aesar™
J60102K2
1L Each for $51.04
 
AAJ60102K3 missing translation for 'viewDocuments' Alfa Aesar™
J60102K3
2L Each for $89.30
 
AAJ60102K7 missing translation for 'viewDocuments' Alfa Aesar™
J60102K7
4L Each for $165.01
 
Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Liquid
1L
C4H11NO3
10X
C(C(CO)(CO)N)O
121.136
CHEBI:9754
8.0
TRIS-buffered saline, low salt
Safety and Handling

Safety and Handling

TSCA : TSCA