Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS-buffered saline (10X, pH 7.6), low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67508AK

 View more versions of this product

Catalog No. AAJ67508AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
MDL Number MFCD00132476
Chemical Name or Material TRIS-buffered saline (10X, pH 7.6)
Color Colorless
Quantity 250mL
TSCA TSCA
Name Note low endotoxin

Specifications

Chemical Name or Material TRIS-buffered saline (10X, pH 7.6)
Name Note low endotoxin
MDL Number MFCD00132476