missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP1759500

 View more versions of this product

Catalog No. BP1759-500


 

Specifications Description & Specifications

Specifications

Tris Buffer
77-86-1
Molecular Biology Grade
1.95 to 2.05M
DNase free
Pass Test
0.0001% max.
Poly Bottle
Protease free
500mL
2M Solution
0.0005% max.
Liquid
0.0001% max.
15, 9951
0.0001% max.
0.0005% max.
0.05 AU max.
Clear

Chemical Identifiers

77-86-1
Tris Buffer, 2M Solution
Molecular Biology Grade

Safety and Handling

SDS & Safety Documents

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:

Signal Word:CAUTION!

Recommended Storage : RT