Please login to your online account to display your discounted pricing

Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP1759100

 View more versions of this product

Catalog No. BP1759-100

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 100mL
Merck Index 15, 9951
DNase DNase free
Protease Protease free
Physical Form Liquid
Concentration 1.95 to 2.05M
Color Clear
Copper (Cu) 0.0001% max.
Magnesium (Mg) 0.0005% max.
Lead (Pb) 0.0001% max.
Optical Density 0.05 AU max.
Chemical Name or Material Tris Buffer
CAS 77-86-1
Name Note 2M Solution
Iron (Fe) 0.0001% max.
Packaging Poly Bottle
Arsenic (As) 0.0005% max.
Identification Pass Test
ChemAlert Storage Symbol Gray
Recommended Storage RT
Health Hazard 1 Signal Word:CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:

Specifications

Chemical Name or Material Tris Buffer
Name Note 2M Solution
CAS 77-86-1
Purity Grade Molecular Biology Grade
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:

RT

SDS & Safety Documents