missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents
$63.20 - $79.59

Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP1759-100
|
Fisher BioReagents
BP1759100 |
100mL | Poly Bottle |
Each for $63.20
|
|
BP1759-500
|
Fisher BioReagents
BP1759500 |
500mL | Poly Bottle |
Each for $79.59
|
|
Description
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers.Chemical Identifiers
77-86-1 | |
121.136 | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
Specifications
1.95 to 2.05M | |
0.0005% max. | |
0.0001% max. | |
DNase free | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
Liquid | |
0.0001% max. | |
0.0005% max. | |
0.05 AU max. | |
Undesignated | |
Tris Buffer, 2M Solution |
77-86-1 | |
C4H11NO3 | |
15, 9951 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
Pass Test | |
Molecular Biology | |
0.0001% max. | |
Poly Bottle | |
Protease free | |
100mL |
Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 1
Flammability:
Signal Word: CAUTION!
Recommended Storage : RT