missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris Buffer, 2M Solution, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 612025000
Specifications
Tris buffer | |
2M solution | |
0.0005% max. | |
MFCD00004679 | |
DNase free | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
0.0001% max. | |
0.0005% max. | |
Protease free | |
500mL |
1.95 to 2.05M | |
77-86-1 | |
C4H11NO3 | |
0.0001% max. | |
Tris(hydroxymethyl)aminomethane | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
Pass Test | |
0.0001% max. | |
0.05 AU max. | |
10.1 to 11.1 |
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
C4H11NO3 | |
MFCD00004679 | |
Tris(hydroxymethyl)aminomethane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |