missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Buffer, 2M Solution, ACROS Organics™

$95.94 - $95.94

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC612021000
SDS View Documents
Fisher Scientific Acros
612021000
100mL
Each for $95.94
 
AC612025000
SDS View Documents
Fisher Scientific Acros
612025000
500mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
Tris(hydroxymethyl)aminomethane
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

1.95 to 2.05M
0.0005% max.
MFCD00004679
DNase free
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
0.0001% max.
0.0005% max.
Protease free
100mL
77-86-1
C4H11NO3
0.0001% max.
Tris(hydroxymethyl)aminomethane
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Pass Test
0.0001% max.
0.05 AU max.
10.1 to 11.1
Tris buffer, 2M solution
SDS