Please login to your online account to display your discounted pricing

Tris Buffer, 100mM, pH 7.4, Molecular Biology Grade, Calbiochem™, EMD Millipore

Useful in the pH range of 10.2 to 10.6

$81.64 - $81.64

Chemical Identifiers

Formula Weight 121.1
Boiling Point 219° to 220°C
CAS 77-86-1
Chemical Name or Material Tris Buffer
Molecular Formula C4H11NO3
Melting Point 168° to 172°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

648315100ML

 
emd millipore corporation
648315100ML
Each for $81.64
Description & Specifications

Specifications

Formula Weight 121.1
Melting Point 168° to 172°C
Boiling Point 219° to 220°C
Quantity 100mL
Purity Molecular Biology Grade
CAS 77-86-1
Chemical Name or Material Tris Buffer
DNase None Detected
RNase None Detected
Molecular Formula C4H11NO3
Synonym tris(Hydroxymethyl)aminomethane, HCl
Solubility Information Soluble in water (800g/L at 20°C)

Widely used biological buffer, Suitable for DNA and RNA applications.