Please login to your online account to display your discounted pricing

Tris Buffer, 1.0 M, pH 8.0, Molecular Biology Grade, Calbiochem™, EMD Millipore

Useful in the pH range of 7.0 to 9.0

$69.82 - $69.82

Chemical Identifiers

Formula Weight 121.1
CAS 77-86-1
Chemical Name or Material Tris Buffer
Concentration or Composition (by Analyte or Components) 1M Solution
Molecular Formula C4H11NO3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

648314100ML

 
emd millipore corporation
648314100ML
Each for $69.82
Description & Specifications

Specifications

Formula Weight 121.1
Quantity 100mL
Molecular Formula C4H11NO3
Recommended Storage 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
RNase None Detected
Chemical Name or Material Tris Buffer
CAS 77-86-1
DNase None Detected
Synonym 1,3-propanediol, 2-amino-2-(hydroxymethyl)-
Concentration or Composition (by Analyte or Components) 1M Solution
Protease None Detected
Purity Molecular Biology Grade

Sterile solution, Suitable for DNA and RNA applications

Safety and Handling

15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.