Please login to your online account to display your discounted pricing

Tris Buffer, 0.3M Solution (Molecular Biology), Fisher BioReagents


Quantity 1L
Packaging Nalgene Poly Bottle

Manufacturer: fisher scientific company  FLBP17611

 View more versions of this product

Catalog No. BP1761-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 1L
Merck Index 15, 9951
Physical Form Liquid
Color Clear
pH 10.1 to 11.1
Protease Protease free
Concentration 0.29 to 0.31M
DNase DNase free
CAS 60-00-4
CAS 77-86-1
Packaging Nalgene Poly Bottle
Name Note 0.3M Solution
ChemAlert Storage Symbol Gray
Recommended Storage RT
Chemical Name or Material Tris Buffer
CAS 7732-18-5
Identification Pass Test

Specifications

Chemical Name or Material Tris Buffer
Name Note 0.3M Solution
CAS 7732-18-5
CAS 77-86-1
CAS 60-00-4
Purity Grade Molecular Biology Grade
Accessories
 

RT

SDS & Safety Documents