Please login to your online account to display your discounted pricing

Tris Buffer, 0.3M Solution (Molecular Biology), Fisher BioReagents


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP1761500

 View more versions of this product

Catalog No. BP1761-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 500mL
Physical Form Liquid
pH 10.1 to 11.1
Color Clear
Protease Protease free
Concentration 0.29 to 0.31M
DNase DNase free
Merck Index 15, 9951
Identification Pass Test
CAS 77-86-1
CAS 60-00-4
CAS 7732-18-5
Name Note 0.3M Solution
ChemAlert Storage Symbol Gray
Chemical Name or Material Tris Buffer
Packaging Poly Bottle
Recommended Storage RT

Specifications

Chemical Name or Material Tris Buffer
Name Note 0.3M Solution
CAS 7732-18-5
CAS 77-86-1
CAS 60-00-4
Purity Grade Molecular Biology Grade
Accessories
 

RT

SDS & Safety Documents