Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris-Borate-EDTA Large Precast Gel, 10%, 16x20cm, 18.3x20cm (back), 1mm thick, 10 well, for Bio-Rad™ systems


Manufacturer: alfa aesar™  J67548KZ

 View more versions of this product

Catalog No. AAJ67548KZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 6 Each
Boiling Point 100°C
Product Type Tris-Borate-EDTA Large Precast Gel
Storage Requirements 2° to 8°C
Color Colorless
Odor Odorless
Format Solid
For Use With (Application) Used for high resolution of proteins or DNA fragments from 10-3,000 base pairs under non-denaturing conditions.