Please login to your online account to display your discounted pricing

TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents


Quantity 4L
Packaging Poly CUBE

Manufacturer: fisher scientific company  BP24304

 View more versions of this product

Catalog No. BP24304

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Electrophoresis Grade
Quantity 4L
Synonym TBE
pH 8.2 to 8.4
Physical Form Liquid
Filtered Through Filtered through a 0.2-micron filter.
Color Colorless
DNase DNase free
Protease Protease free
CAS 7732-18-5
Name Note 1X Solution
CAS 77-86-1
ChemAlert Storage Symbol Gray
Electrophoresis Pass Test
CAS 6381-92-6
Packaging Poly CUBE
Recommended Storage RT
Chemical Name or Material Tris-Borate-EDTA
CAS 10043-35-3

Specifications

Chemical Name or Material Tris-Borate-EDTA
Name Note 1X Solution
CAS 77-86-1
CAS 6381-92-6
CAS 10043-35-3
CAS 7732-18-5
Purity Grade Electrophoresis Grade

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.089M tris, 0.089M boric acid, and 0.002M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only

Accessories
 

RT

SDS & Safety Documents