Please login to your online account to display your discounted pricing

TRIS (Base), BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

High quality biochemicals for research use

$744.31 - $744.31

Chemical Identifiers

Chemical Name or Material TRIS (Base)
CAS 77-86-1
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Molecular Formula NH2C(CH2OH)3
Formula Weight 121.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004510

 
avantor performance
409906
Poly Pail 5kg Each for $744.31
Description & Specifications

Specifications

Formula Weight 121.14
CAS 77-86-1
Molecular Formula NH2C(CH2OH)3
Packaging Poly Pail
Purity Grade BAKER ANALYZED™ Biochemical Reagent
Quantity 5kg
Chemical Name or Material TRIS (Base)

    For Liquid Chromatography Applications