Please login to your online account to display your discounted pricing

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
CAS 80049-61-2
Assay ≥97.5% (ex Ru)
Assay Percent Range 98%
Molecular Formula C11H14F6N3PRu
Formula Weight 434.29
MDL Number MFCD02684569
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377892500

  SDS
thermo fisher scientific
AC377892500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC377890010

  SDS
thermo fisher scientific
AC377890010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
CAS 80049-61-2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Physical Form Powder
Assay ≥97.5% (ex Ru)
Assay Percent Range 98%
Molecular Formula C11H14F6N3PRu
Packaging Glass bottle
Formula Weight 434.29
MDL Number MFCD02684569
Color Yellow to orange or brown
Quantity 250mg
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.