Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate

Chemical Identifiers

Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
CAS 80049-61-2
Linear Formula [(C5H5)Ru(CH3CN)3]PF6
Formula Weight 434.28
MDL Number MFCD02684569
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47214MD

  SDS
alfa aesar™
47214MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4721403

  SDS
alfa aesar™
4721403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
CAS 80049-61-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Physical Form Powder
Linear Formula [(C5H5)Ru(CH3CN)3]PF6
Formula Weight 434.28
MDL Number MFCD02684569
Color Yellow to orange
Quantity 250mg
Sensitivity Air & Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C