Please login to your online account to display your discounted pricing

TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents

$79.05 - $915.56

Chemical Identifiers

Chemical Name or Material Tris-Acetate-EDTA
Name Note 50X Solution
CAS 7732-18-5
CAS 77-86-1
CAS 60-00-4
CAS 6850-28-8
Purity Grade Electrophoresis Grade
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Liquid
Synonym TAE
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP13324

  SDS
fisher scientific company
BP13324
Poly CUBE 4L Each for $258.63

BP1332-500

  SDS
fisher scientific company
FLBP1332500
Poly Bottle 500mL Each for $79.05
Case of 6 Each for $474.30

BP1332-1

  SDS
fisher scientific company
FLBP13321
Nalgene Poly Bottle 1L Each for $107.02
Case of 6 Each for $642.12

BP1332-20

  SDS
fisher scientific company
FLBP133220
Poly CUBE 20L Each for $915.56
Description & Specifications

Specifications

Purity Grade Electrophoresis Grade
Quantity 4L
Filtered Through Filtered through a 0.2-micron filter.
Physical Form Liquid
Conductivity [NULL]
DNase DNase free
Protease Protease free
Chemical Name or Material Tris-Acetate-EDTA
CAS 6850-28-8
Purity Grade Notes DNase-, RNase- and Protease-Free
CAS 7732-18-5
ChemAlert Storage Symbol Gray
Electrophoresis Pass Test
Concentration or Composition (by Analyte or Components) Tris (24%), Acetic Acid (5.0%), and EDTA (<2%).
CAS 77-86-1
Packaging Poly CUBE
CAS 60-00-4
Name Note 50X Solution
Synonym TAE
Recommended Storage RT

Tris–Acetate–EDTA (TAE) is commonly used as a buffer for nucleic acid electrophoresis.

Safety and Handling
 

RT