missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT
Supplier: Fisher BioReagents BP13324
Description
Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT
- Save time and simplify your buffer preparation step by using Fisher BioReagents 50X TAE Solution - simply dilute as needed
- 50X solution contains 2M tris-acetate and 0.050M EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 50X TAE
- Solution with 49 volumes of ultrapure water, such as BP2819
- Tested for the absence of DNase, RNase, and protease
- Filtered, autoclaved, and functionally tested for use in electrophoresis applications
- TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
- Intended for laboratory research use only
Specifications
Tris-Acetate-EDTA Buffer | |
Gray | |
50X | |
4 L | |
Filtered through a 0.2-micron filter. | |
Tris (24%), Acetic Acid (5.0%), and EDTA (<2%) | |
Protease free | |
Electrophoresis |
50X Solution | |
77-86-1,60-00-4,6850-28-8 | |
Liquid | |
DNase free | |
DNase-, RNase- and Protease-Free | |
Poly CUBE | |
TAE | |
Tris (24%), Acetic Acid (5.0%), and EDTA (<2%). |