Please login to your online account to display your discounted pricing

TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents


Quantity 4L
Packaging Poly CUBE

Manufacturer: fisher scientific company  BP24344

 View more versions of this product

Catalog No. BP24344

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Electrophoresis Grade
Quantity 4L
Synonym TAE
Filtered Through Filtered through a 0.2-micron filter.
Color Clear
Physical Form Liquid
DNase DNase free
Protease Protease free
CAS 10378-23-1
Name Note 1X Solution
CAS 77-86-1
ChemAlert Storage Symbol Gray
Electrophoresis Pass Test
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0
Chemical Name or Material Tris-Acetate-EDTA
CAS 7732-18-5
Health Hazard 2 CAUTION!
Packaging Poly CUBE
CAS 6850-28-8
Recommended Storage RT
CAS 139-33-3

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 1X Solution
CAS 77-86-1
CAS 6850-28-8
CAS 139-33-3
CAS 10378-23-1
CAS 7732-18-5
Purity Grade Electrophoresis Grade

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TAE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.04M tris-acetate and 0.001M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only

Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

RT

SDS & Safety Documents