TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Manufacturer:  Fisher Scientific Company BP243420
 View more versions of this product
Catalog No. BP2434-20
Specifications
Description
Tris-Acetate-EDTA; Electrophoresis; Fisher BioReagents; 20L; Poly CUBE; CAS: 77-86-1
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TAE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.04M tris-acetate and 0.001M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
Description & Specifications
Specifications
Tris-Acetate-EDTA
77-86-1,6850-28-8,139-33-3,10378-23-1,7732-18-5
Liquid
Pass Test
Poly CUBE
Filtered through a 0.2-micron filter.
20L
1X Solution
Electrophoresis Grade
DNase free
TAE
Protease free
Clear
Chemical Identifiers
77-86-1,6850-28-8,139-33-3,10378-23-1,7732-18-5
Tris-Acetate-EDTA, 1X Solution
Electrophoresis Grade
Safety and Handling
SDS & Safety Documents
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT