missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP13354

 View more versions of this product

Catalog No. BP13354


 

Specifications

Description

Tris-Acetate-EDTA; Electrophoresis-Grade; Colorless Liquid; Fisher BioReagents; 4L; Poly CUBE; CAS: 64-19-7

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
  • 10X solution contains 0.4M tris-acetate and 0.01M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
Description & Specifications

Specifications

Electrophoresis Grade
Liquid
DNase free
TAE
DNase-, RNase- and Protease-Free
7732-18-5,64-19-7,77-86-1,60-00-4
Pass Test
4L
Protease free
Filtered through a 0.2-micron filter.
Poly CUBE
Tris-Acetate-EDTA
Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
10X Solution

Chemical Identifiers

64-19-7,7732-18-5,60-00-4,77-86-1
TAE
Electrophoresis Grade, DNase-, RNase- and Protease-Free
Liquid
Tris-Acetate-EDTA, 10X Solution

Safety and Handling

SDS & Safety Documents

Recommended Storage : RT