Please login to your online account to display your discounted pricing

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents


Quantity 1L
Packaging Nalgene Poly Bottle

Manufacturer: fisher scientific company  FLBP13351

 View more versions of this product

Catalog No. BP1335-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Electrophoresis Grade
Quantity 1L
Filtered Through Filtered through a 0.2-micron filter.
Physical Form Liquid
Conductivity [NULL]
DNase DNase free
Protease Protease free
CAS 64-19-7
Synonym TAE
Concentration or Composition (by Analyte or Components) Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
Packaging Nalgene Poly Bottle
Recommended Storage RT
Chemical Name or Material Tris-Acetate-EDTA
CAS 77-86-1
Purity Grade Notes DNase-, RNase- and Protease-Free
ChemAlert Storage Symbol Gray
Electrophoresis Pass Test
CAS 7732-18-5
Name Note 10X Solution
CAS 60-00-4

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 10X Solution
CAS 64-19-7
CAS 7732-18-5
CAS 60-00-4
CAS 77-86-1
Purity Grade Electrophoresis Grade
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Liquid
Synonym TAE

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
  • 10X solution contains 0.4M tris-acetate and 0.01M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only

Accessories
 

RT

SDS & Safety Documents