missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure, Thermo Scientific™

Manufacturer:  Thermo Scientific™ J75825A7

 View more versions of this product

Catalog No. AAJ75825A7



Specifications

Specifications

Tris
C4H11NO3
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Molecular Biology
219°C to 220°C
5kg
77-86-1
NH2C(CH2OH)3
Tris(hydroxymethyl)amino methane
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Crystalline Powder
Plastic bottle
171°C to 172°C

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
Tris(hydroxymethyl)amino methane
CHEBI:9754
C(C(CO)(CO)N)O
Safety and Handling

Safety and Handling

TSCA : Y

Documents