missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(8-hydroxyquinolinato)aluminum, 99%

Manufacturer:  Alfa Aesar™ H5565614

 View more versions of this product

Catalog No. AAH5565614



Description

Description

Applications
Key green-light emitting and electron transport material for OLEDs.

Solubility
Solubility in chloroform

Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

Tris(8-hydroxyquinolinato)aluminum
>300°C
2085-33-8
14,370
459.44
99%
Yellow to Green
25g
MFCD00191693
aluminium tris quinolin-8-olate
Powder

Chemical Identifiers

2085-33-8
MFCD00191693
aluminium tris quinolin-8-olate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-227-0

RTECSNumber : BD2231000

TSCA : No

Recommended Storage : Ambient temperatures

Documents