Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III), 99%

$98.31 - $98.31

Chemical Identifiers

Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III)
CAS 17978-77-7
Molecular Formula C30H30F21O6Pr
Formula Weight 1026.45
MDL Number MFCD00064656
Synonym Praseodymium-fod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1416603

 
alfa aesar™
A1416603
1g Each for $98.31
Description & Specifications

Specifications

Formula Weight 1026.45
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III)
TSCA TSCA
Assay 99%
Molecular Formula C30H30F21O6Pr
MDL Number MFCD00064656
CAS 17978-77-7
Synonym Praseodymium-fod
Melting Point 215° to 219°C
Quantity 1g

Applications
Lewis catalyst for aldol reactions. NMR shift reagent.

Solubility
Soluble in hexane, methanol, tetrahydrofuran.

Notes
NMR Shift Reagent. Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.