Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III), 98+%

$58.81 - $58.81

Chemical Identifiers

Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III)
CAS 18323-98-3
Molecular Formula C30H30DyF21O6
Formula Weight 1048.04
MDL Number MFCD00010463
Synonym Dysprosium-fod
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2175903

 
alfa aesar™
B2175903
1g Each for $58.81
Description & Specifications

Specifications

Formula Weight 1048.04
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III)
Assay ≥98%
Molecular Formula C30H30DyF21O6
MDL Number MFCD00010463
CAS 18323-98-3
Synonym Dysprosium-fod
Melting Point 184° to 186°C
Quantity 1g