missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(4-trifluoromethylphenyl)phosphine, 99%, ACROS Organics™

Chemical Identifiers

CAS 13406-29-6
Molecular Formula C21H12F9P
Molecular Weight (g/mol) 466.29
MDL Number MFCD00058883
InChI Key PXYCJKZSCDFXLR-UHFFFAOYSA-N
Synonym acmc-20ajbq, phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl, phosphine, tris 4-trifluoromethyl phenyl, tris 4-trifluoromethyl phenyl phosphane, tris 4-trifluoromethyl phenyl phosphine, tris 4-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine,min
PubChem CID 139448
IUPAC Name tris[4-(trifluoromethyl)phenyl]phosphane
SMILES C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC355340050
SDS View Documents
Thermo Fisher Scientific
AC355340050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

13406-29-6
466.29
PXYCJKZSCDFXLR-UHFFFAOYSA-N
139448
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F
C21H12F9P
MFCD00058883
acmc-20ajbq, phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl, phosphine, tris 4-trifluoromethyl phenyl, tris 4-trifluoromethyl phenyl phosphane, tris 4-trifluoromethyl phenyl phosphine, tris 4-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine,min
tris[4-(trifluoromethyl)phenyl]phosphane

Specifications

Authentic
Undesignated
5g
98.5% min. (GC)
MFCD00058883
Solubility in water: insoluble.
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F
466.29
466.29
99%
Glass bottle
72.0°C to 75.0°C
13406-29-6
C21H12F9P
acmc-20ajbq, phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl, phosphine, tris 4-trifluoromethyl phenyl, tris 4-trifluoromethyl phenyl phosphane, tris 4-trifluoromethyl phenyl phosphine, tris 4-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine, tris p-trifluoromethylphenyl phosphine,min
PXYCJKZSCDFXLR-UHFFFAOYSA-N
tris[4-(trifluoromethyl)phenyl]phosphane
139448
Liquid
Tris(4-trifluoromethylphenyl)phosphine
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning