Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris[4-(trifluoromethyl)phenyl]phosphine, 98%

$180.09 - $691.14

Chemical Identifiers

Chemical Name or Material Tris[4-(trifluoromethyl)phenyl]phosphine
CAS 13406-29-6
Molecular Formula C21H12F9P
Formula Weight 466.29
MDL Number MFCD00058883
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1288503

 
alfa aesar™
A1288503
1g Each for $180.09

AAA1288506

 
alfa aesar™
A1288506
5g Each for $691.14
Description & Specifications

Specifications

Formula Weight 466.29
Chemical Name or Material Tris[4-(trifluoromethyl)phenyl]phosphine
Assay 98%
Molecular Formula C21H12F9P
MDL Number MFCD00058883
CAS 13406-29-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 72° to 75°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Tris[4-(trifluoromethyl)phenyl]phosphine is a raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.